Learn More
4,4'-Azo-bis(4-cyanovaleric acid), ∽98%, MP Biomedicals™
Product Code. 11433271 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11433271 25 g 25g
11423271 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11433271 Supplier MP Biomedicals Supplier No. 0215041225
 Restricted Item
Estimated Shipment: 18-11-2026
Chemical Identifiers
CAS 2638-94-0
Molecular Formula C12H16N4O4
Molecular Weight (g/mol) 280.284
MDL Number MFCD00002799
InChI Key VFXXTYGQYWRHJP-UHFFFAOYSA-N
Synonym 4,4'-azobis 4-cyanovaleric acid, azobis cyanovaleric acid, 4,4'-diazene-1,2-diyl bis 4-cyanopentanoic acid, 4,4'-azobis 4-cyanopentanoic acid, acva, 4,4-azobis 4-cyanovaleric acid, valeric acid, 4,4'-azobis 4-cyano, pentanoic acid, 4,4'-azobis 4-cyano, 4,4'-azobis 4-cyano valeric acid acva
PubChem CID 92938
IUPAC Name 4-[(4-carboxy-2-cyanobutan-2-yl)diazenyl]-4-cyanopentanoic acid
SMILES CC(CCC(=O)O)(C#N)N=NC(C)(CCC(=O)O)C#N
Specifications
Color White
Assay Percent Range ∽ 98%
Quantity 25 g
Formula Weight 280.28
Percent Purity 98%
Grade Research
Physical Form Powder
Chemical Name or Material 4, 4'-Azo-bis(4-cyanovaleric acid), Polymer synthesis