missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4,4'-Azo-bis(4-cyanovaleric acid), ∽98%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Specifications
Specifications
| Color | White |
| Assay Percent Range | ∽ 98% |
| Quantity | 5 g |
| Formula Weight | 280.28 |
| Physical Form | Powder |
| Chemical Name or Material | 4, 4'-Azo-bis(4-cyanovaleric acid), Polymer synthesis |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction