Learn More
4,4,5,5-Tetramethyl-2-phenyl-1,3,2-dioxaborolane, 95%, BLD Pharm™
Product Code. 30947712
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
100 g
500 g
1 kg
Unit Size:
100g
10g
1kg
25g
500g
5g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30947712 500 g 500g
30942106 5 g 5g
30935893 25 g 25g
30941102 10 g 10g
30941566 100 g 100g
30934121 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 30947712 Supplier BLD Pharm Supplier No. BD14170500g
 Restricted Item
Estimated Shipment: 27-11-2026
Spezifikation
CAS 24388-23-6
Molecular Formula C12H17BO2
MDL Number MFCD00966985
Quantity 500 g
SMILES CC1(C)C(C)(C)OB(C2=CC=CC=C2)O1
Molecular Weight (g/mol) 204.07
Percent Purity 95%
Chemical Name or Material 4,4,5,5-Tetramethyl-2-phenyl-1,3,2-dioxaborolane