missing translation for 'onlineSavingsMsg'
Learn More

4,4,4-Trifluorocrotonic acid, 97%, Thermo Scientific™

Product Code. 15525926 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15525926

Brand: Thermo Scientific Acros 456510050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 71027-02-6
Molecular Formula C4H3F3O2
Molecular Weight (g/mol) 140.06
InChI Key QZBAYURFHCTXOJ-OWOJBTEDSA-N
Synonym 4,4,4-trifluorocrotonic acid, e-4,4,4-trifluorobut-2-enoic acid, 2e-4,4,4-trifluorobut-2-enoic acid, 4,4,4,-trifluorobut-2-enoic acid, 4,4,4-trifluoro-2-butenoic acid, hiddpndiaicichibejifaap, trifluorocrotonic acid, pubchem22644, 4,4,4-trifluoro crotonic acid
PubChem CID 5709614
IUPAC Name (E)-4,4,4-trifluorobut-2-enoic acid
SMILES C(=CC(F)(F)F)C(=O)O

Specifications

Melting Point 53.0°C to 58.0°C
Boiling Point 68.0°C to 69.0°C (20.0 mmHg)
Flash Point >110°C
Infrared Spectrum Authentic
Quantity 5 g
Formula Weight 140.06
Percent Purity 97%
Chemical Name or Material 4,4,4-Trifluorocrotonic acid
compliance-icons
Product Identifier
  • 4,4,4-Trifluorocrotonic acid, 97%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.