accessibility menu, dialog, popup

UserName

CAS RN 71027-02-6

FFFOOH(E/Z)

CAS RN 71027-02-6

IUPAC Name: 4,4,4-trifluorobut-2-enoic acid
Synonyms: 4,4,4-trifluorobut-2-enoic acid
Molecular Weight (g/mol): 140.06
Molecular Formula: C4H3F3O2
InChi Key: QZBAYURFHCTXOJ-UHFFFAOYSA-N
SMILES: OC(=O)C=CC(F)(F)F
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results