missing translation for 'onlineSavingsMsg'
Learn More

4,4'-[2,2,2-Trifluoro-1-(trifluoromethyl)ethylidene]-O-benzyl Mono-Beta-D-glucuroniC Acid Methyl Ester 2,3,4-O-Triacetate, TRC

Product Code. 30456293
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456293 5 mg 5mg
30454706 10 mg 10mg
30451130 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456293 Supplier Toronto Research Chemicals Supplier No. TRCT7927655MG
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4,4'-[2,2,2-Trifluoro-1-(trifluoromethyl)ethylidene]-O-benzyl Mono-beta-D-glucuroniC Acid Methyl Ester 2,3,4-O-Triacetate
Analyte or Component Names 4,4'-[2,2,2-Trifluoro-1-(trifluoromethyl)ethylidene]-O-benzyl Mono-beta-D-glucuroniC Acid Methyl Ester 2,3,4-O-Triacetate
Shipping Condition Room Temperature
Molecular Formula C35H32F6O11
InChI Formula InChI=1S/C35H32F6O11/c1-19(42)48-27-28(49-20(2)43)30(50-21(3)44)32(52-29(27)31(45)46-4)51-26-16-12-24(13-17-26)33(34(36,37)38,35(39,40)41)23-10-14-25(15-11-23)47-18-22-8-6-5-7-9-22/h5-17,27-30,32H,18H2,1-4H3/t27?,28?,29?,30?,32-/m1/s1
Solution Type Neat
SMILES FC(F)(F)C(C1=CC=C(OCC2=CC=CC=C2)C=C1)(C(F)(F)F)C3=CC=C(O[C@H]4[C@H](OC(C)=O)[C@@H](OC(C)=O)[C@H](OC(C)=O)[C@@H](C(OC)=O)O4)C=C3
IUPAC Name (2S,3R,4S,5S,6S)-2-(4-(2-(4-(benzyloxy)phenyl)-1,1,1,3,3,3-hexafluoropropan-2-yl)phenoxy)-6-(methoxycarbonyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 742.61
Quantity 5 mg
Formula Weight 742.18
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.