Learn More
4-[(3-Methylphenyl)amino]-3-pyridinesulfonamide, TRC
Specifications
Specifications
| Chemical Name or Material | 4-[(3-Methylphenyl)amino]pyridine-3-sulfonamide |
| CAS | 72811-73-5 |
| Molecular Formula | C12 H13 N3 O2 S |
| Quantity | 500 mg |
| Synonym | 4-[(3-Methylphenyl)amino]pyridine-3-sulfonamide, Torasemide','Anhydrous Imp. B (EP) |
| InChI Formula | InChI=1S/C12H13N3O2S/c1-9-3-2-4-10(7-9)15-11-5-6-14-8-12(11)18(13,16)17/h2-8H,1H3,(H,14,15)(H2,13,16,17) |
| SMILES | Cc1cccc(Nc2ccncc2S(=O)(=O)N)c1 |
| IUPAC Name | 4-(3-methylanilino)pyridine-3-sulfonamide |
| Molecular Weight (g/mol) | 263.32 |
| Formula Weight | 263.0728 |