Learn More
4-(3-Chloro-4-fluorophenylamino)-7-fluoro-6-nitroquinazoline, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(3-Chloro-4-fluorophenylamino)-7-fluoro-6-nitroquinazoline |
| CAS | 162012-67-1 |
| Molecular Formula | C14H7ClF2N4O2 |
| Quantity | 100 mg |
| Synonym | N-(3-Chloro-4-fluorophenyl)-7-fluoro-6-nitro-4-quinazolinamine, (3-Chloro-4-fluorophenyl)(7-fluoro-6-nitroquinazolin-4-yl)amine |
| InChI Formula | InChI=1S/C14H7ClF2N4O2/c15-9-3-7(1-2-10(9)16)20-14-8-4-13(21(22)23)11(17)5-12(8)18-6-19-14/h1-6H,(H,18,19,20) |
| SMILES | [O-][N+](=O)c1cc2c(Nc3ccc(F)c(Cl)c3)ncnc2cc1F |
| IUPAC Name | N-(3-chloro-4-fluorophenyl)-7-fluoro-6-nitroquinazolin-4-amine |
| Molecular Weight (g/mol) | 336.68 |