missing translation for 'onlineSavingsMsg'
Learn More

4-(3-Acetoxypropyl)phenyl Acetate, TRC

Product Code. 30312675
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30312675 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312675 Supplier Toronto Research Chemicals Supplier No. TRCA167695100MG
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 4-(3-Acetoxypropyl)phenyl Acetate
CAS 25823-43-2
Molecular Formula C13H16O4
Quantity 100 mg
Synonym 4-(Acetyloxy)benzenepropanol Acetate, 3-(p-Hydroxyphenyl)-1-propanol Diacetate
InChI Formula InChI=1S/C13H16O4/c1-10(14)16-9-3-4-12-5-7-13(8-6-12)17-11(2)15/h5-8H,3-4,9H2,1-2H3
SMILES CC(OCCCC1=CC=C(OC(C)=O)C=C1)=O
IUPAC Name 3-(4-acetyloxyphenyl)propyl acetate
Molecular Weight (g/mol) 236.26
Formula Weight 236.1
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.