missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Methyl-1H-imidazol-1-yl)benzylamine, 90%, Thermo Scientific™

Catalog Number 10508863
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
883291-45-0
C11H13N3
187.246
MFCD06617944
JOMOQABDRBJMDQ-UHFFFAOYSA-N
4-2-methyl-1h-imidazol-1-yl benzylamine, 1-4-2-methyl-1h-imidazol-1-yl phenyl methanamine, 4-2-methylimidazol-1-yl phenyl methanamine, benzenemethanamine,4-2-methyl-1h-imidazol-1-yl, 4-2-methyl-1h-imidazol-1-yl phenyl methanamine, 4-2-methylimidazolyl phenyl methylamine, 1-4-2-methylimidazol-1-yl phenyl methanamine
4913134
[4-(2-methylimidazol-1-yl)phenyl]methanamine
CC1=NC=CN1C2=CC=C(C=C2)CN
This item is not returnable. View return policy
883291-45-0
C11H13N3
187.246
MFCD06617944
JOMOQABDRBJMDQ-UHFFFAOYSA-N
4-2-methyl-1h-imidazol-1-yl benzylamine, 1-4-2-methyl-1h-imidazol-1-yl phenyl methanamine, 4-2-methylimidazol-1-yl phenyl methanamine, benzenemethanamine,4-2-methyl-1h-imidazol-1-yl, 4-2-methyl-1h-imidazol-1-yl phenyl methanamine, 4-2-methylimidazolyl phenyl methylamine, 1-4-2-methylimidazol-1-yl phenyl methanamine
4913134
[4-(2-methylimidazol-1-yl)phenyl]methanamine
CC1=NC=CN1C2=CC=C(C=C2)CN
CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 187.24
Percent Purity 90%
Chemical Name or Material 4-(2-Methyl-1H-imidazol-1-yl)benzylamine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'