missing translation for 'onlineSavingsMsg'
Learn More

4-(2-Aminoethyl)tetrahydropyran, 97%, Thermo Scientific™

Product Code. 15528741 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15528741 1 g 1g
15538741 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15528741 Supplier Thermo Scientific Acros Supplier No. 455860010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 65412-03-5
Molecular Formula C7H16NO
Molecular Weight (g/mol) 130.21
MDL Number MFCD02179433
InChI Key BZMADPOGYCRPAI-UHFFFAOYSA-O
Synonym 4-2-aminoethyl tetrahydropyran, 4-2-aminoethyl tetrahydro-2h-pyran, 2-oxan-4-yl ethan-1-amine, 2-oxan-4-yl ethanamine, 2-tetrahydro-2h-pyran-4-yl ethanamine, 4-aminoethyl-tetrahydropyrane, 2h-pyran-4-ethanamine, tetrahydro, 4-aminoethyl tetrahydropyran, 2-tetrahydro-pyran-4-yl-ethylamine, 2-tetrahydro-pyran-4-yl ethylamine
PubChem CID 2773198
SMILES [NH3+]CCC1CCOCC1

Specifications

Boiling Point 88.0°C to 89.0°C (13.0 mmHg)
Flash Point 74°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
IUPAC Name 2-(oxan-4-yl)ethanamine
Formula Weight 129.2
Percent Purity 97%
Chemical Name or Material 4-(2-Aminoethyl)tetrahydropyran
compliance-icons
Product Identifier
  • 4-(2-Aminoethyl)tetrahydropyran, 97%
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.