Learn More
4-(2,4-Difluorophenyl)-3-methyl-2(5H)-furanone, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(2,4-Difluorophenyl)-3-methyl-2(5H)-furanone |
| CAS | 102748-42-4 |
| Molecular Formula | C11 H8 F2 O2 |
| Quantity | 500 mg |
| InChI Formula | InChI=1S/C11H8F2O2/c1-6-9(5-15-11(6)14)8-3-2-7(12)4-10(8)13/h2-4H,5H2,1H3 |
| SMILES | CC1=C(COC1=O)c2ccc(F)cc2F |
| IUPAC Name | 3-(2,4-difluorophenyl)-4-methyl-2H-furan-5-one |
| Molecular Weight (g/mol) | 210.177 |
| Formula Weight | 210.049 |