Learn More
[4-[2-[[(3-Ethyl-2,5-dihydro-4-methyl-2-oxo-1H-pyrrol-1-yl)carbonyl]amino]ethyl]phenyl]sulfonyl]carbamic Acid Ethyl Ester, TRC
Product Code. 17991475
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17991475 10 mg 10mg
17932785 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17991475 Supplier Toronto Research Chemicals Supplier No. TRCE91654010MG
 Restricted Item
Estimated Shipment: 09-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 318515-70-7
Molecular Weight (g/mol) 423.48
IUPAC Name ethyl N-[4-[2-[(4-ethyl-3-methyl-5-oxo-2H-pyrrole-1-carbonyl)amino]ethyl]phenyl]sulfonylcarbamate
SMILES CCOC(=O)NS(=O)(=O)c1ccc(CCNC(=O)N2CC(=C(CC)C2=O)C)cc1
Specifications
Molecular Formula C19 H25 N3 O6 S
Quantity 10 mg
Synonym Carbamic acid, N-[[4-[2-[[(3-ethyl-2,5-dihydro-4-methyl-2-oxo-1H-pyrrol-1-yl)carbonyl]amino]ethyl]phenyl]sulfonyl]-, ethyl ester, Carbamic acid, [[4-[2-[[(3-ethyl-2,5-dihydro-4-methyl-2-oxo-1H-pyrrol-1-yl)carbonyl]amino]ethyl]phenyl]sulfonyl]-, ethyl ester (9CI), Ethyl [[4-[2-[[(3-Ethyl-4-methyl-2-oxo-2,5-dihydro-1H-pyrrol-1-yl)carbonyl]amino]ethyl]phenyl]sulphonyl]carbamate
InChI Formula InChI=1S/C19H25N3O6S/c1-4-16-13(3)12-22(17(16)23)18(24)20-11-10-14-6-8-15(9-7-14)29(26,27)21-19(25)28-5-2/h6-9H,4-5,10-12H2,1-3H3,(H,20,24)(H,21,25)
Formula Weight 423.1464
Percent Purity >95%
Chemical Name or Material Ethyl ([4-[2-[[(3-Ethyl-4-methyl-2-oxo-2,5-dihydro-1H-pyrrol-1-yl)carbonyl]amino)ethyl]phenyl]sulphonyl)carbamate
Grade HPLC