missing translation for 'onlineSavingsMsg'
Learn More

4-(1H-Imidazol-1-ylmethyl)benzonitrile, ≥97%, Thermo Scientific™

Product Code. p-3175377
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10117622 1 g 1g
10672675 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10117622 Supplier Thermo Scientific Maybridge Supplier No. CC43316DA
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 112809-54-8
Molecular Formula C11H9N3
Molecular Weight (g/mol) 183.214
MDL Number MFCD08060525
InChI Key LUSFCTSUDCCYLQ-UHFFFAOYSA-N
Synonym 4-1h-imidazol-1-ylmethyl benzonitrile, 4-imidazol-1-ylmethyl benzonitrile, 1-4-cyanobenzyl imidazole, 4-1h-imidazol-1-yl methyl benzonitrile, 1-4-cyanobenzyl-1h-imidazole, 4-imidazol-1-ylmethyl-benzonitrile, 4-1-imidazolylmethyl-benzonitrile, benzonitrile,4-1h-imidazol-1-ylmethyl, acmc-1cab3
PubChem CID 1236449
IUPAC Name 4-(imidazol-1-ylmethyl)benzonitrile
SMILES C1=CC(=CC=C1CN2C=CN=C2)C#N

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 183.21
Percent Purity 97%
Chemical Name or Material 4-(1H-Imidazol-1-ylmethyl)benzonitrile
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.