accessibility menu, dialog, popup

UserName

CAS RN 112809-54-8

NNN

CAS RN 112809-54-8

IUPAC Name: 4-[(1H-imidazol-1-yl)methyl]benzonitrile
Synonyms: 4-(imidazol-1-ylmethyl)benzonitrile
Molecular Weight (g/mol): 183.21
Molecular Formula: C11H9N3
InChi Key: LUSFCTSUDCCYLQ-UHFFFAOYSA-N
SMILES: N#CC1=CC=C(CN2C=CN=C2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results