missing translation for 'onlineSavingsMsg'
Learn More

4-(1-Methyl-4-piperidinylmethoxy)aniline, 96%

Product Code. 15469017
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15469017 1 g 1g
15479017 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15469017 Supplier Thermo Scientific Alfa Aesar Supplier No. H52351.03
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 902454-26-6 | C13H20N2O | 220.316 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 902454-26-6
Molecular Formula C13H20N2O
Molecular Weight (g/mol) 220.316
MDL Number MFCD09909628
InChI Key WZRKMUXDLMLRJV-UHFFFAOYSA-N
Synonym 4-1-methylpiperidin-4-yl methoxy aniline, 4-1-methyl-4-piperidinylmethoxy aniline, 4-1-methylpiperidin-4-yl methoxy phenylamine, 4-1-methyl-piperidin-4-ylmethoxy-phenylamine, 4-1-methyl-piperidin-4-ylmethoxy-phenyl amine, 4-1-methyl-4-piperidinyl methoxy benzenamine
PubChem CID 11564798
IUPAC Name 4-[(1-methylpiperidin-4-yl)methoxy]aniline
SMILES CN1CCC(CC1)COC2=CC=C(C=C2)N

Specifications

Quantity 1 g
UN Number UN2811
Formula Weight 220.31
Percent Purity 96%
Chemical Name or Material 4-(1-Methyl-4-piperidinylmethoxy)aniline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.