Learn More
4-(1,1-Dioxidothiomorpholino)benzaldehyde, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 27913-96-8 |
| Molecular Formula | C11H13NO3S |
| MDL Number | MFCD08276340 |
| Quantity | 5 g |
| SMILES | O=CC1=CC=C(N(CC2)CCS2(=O)=O)C=C1 |
| Molecular Weight (g/mol) | 239.29 |
| Percent Purity | 98% |
| Chemical Name or Material | 4-(1,1-Dioxidothiomorpholino)benzaldehyde |