Learn More
4-[[(1,1-Dimethylethyl)dimethylsilyl]oxy]-1-butanamine, TRC
Specifications
Specifications
| Chemical Name or Material | 4-(((1,1-Dimethylethyl)dimethylsilyl)oxy)-1-butanamine |
| CAS | 245660-15-5 |
| Molecular Formula | C10H25NOSi |
| Quantity | 2.5 g |
| Synonym | 4-[(tert-Butyldimethylsilyl)oxy]butan-1-amine, [4-[(tert-Butyldimethylsilyl)oxy]butyl]amine |
| InChI Formula | InChI=1S/C10H25NOSi/c1-10(2,3)13(4,5)12-9-7-6-8-11/h6-9,11H2,1-5H3 |
| SMILES | CC(C)(C)[Si](C)(C)OCCCCN |
| IUPAC Name | 4-[tert-butyl(dimethyl)silyl]oxybutan-1-amine |
| Molecular Weight (g/mol) | 203.4 |
| Formula Weight | 203.1705 |