Learn More
(3S)-N-Methyl-Gamma-(1-naphthalenyloxy)-3-thiophenepropanamine Maleic Acid Salt, TRC
Specifications
Specifications
| Chemical Name or Material | (3S)-N-Methyl-Gamma-(1-naphthalenyloxy)-3-thiophenepropanamine Maleic Acid Salt |
| CAS | 959392-23-5 |
| Molecular Formula | C18 H19 N O S . C4 H4 O4 |
| Quantity | 5 mg |
| InChI Formula | InChI=1S/C18H19NOS.C4H4O4/c1-19-11-9-17(15-10-12-21-13-15)20-18-8-4-6-14-5-2-3-7-16(14)18;5-3(6)1-2-4(7)8/h2-8,10,12-13,17,19H,9,11H2,1H3;1-2H,(H,5,6)(H,7,8)/b;2-1-/t17-;/m0./s1 |
| SMILES | CNCC[C@H](Oc1cccc2ccccc12)c3ccsc3.OC(=O)\C=C/C(=O)O |
| IUPAC Name | (Z)-but-2-enedioic acid;(3S)-N-methyl-3-naphthalen-1-yloxy-3-thiophen-3-ylpropan-1-amine |
| Molecular Weight (g/mol) | 413.487 |
| Formula Weight | 413.13 |