missing translation for 'onlineSavingsMsg'
Learn More

[(3S,4R)-4-(4-Fluorophenyl)-1-methylpiperidin-3-yl]methanol, Mikromol™

Product Code. 30463864
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30463864 4 x 25 mg -
30050604 100 mg 100mg
30050583 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30463864 Supplier Mikromol Supplier No. MM1032.1200254AMP
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

[(3S,4R)-4-(4-Fluorophenyl)-1-methylpiperidin-3-yl]methanol, Mikromol
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Paroxol
CAS 105812-81-5
Analyte or Component Names Paroxol
Shipping Condition Room Temperature
Molecular Formula C13 H18 F N O
Certifications/Compliance ISO 17025
Synonym [(3S,4R)-4-(4-Fluorophenyl)-1-methylpiperidin-3-yl]methanol, BRL-42496, Paroxetine Stage 3B intermediate, BRL 29226AD
InChI Formula InChI=1S/C13H18FNO/c1-15-7-6-13(11(8-15)9-16)10-2-4-12(14)5-3-10/h2-5,11,13,16H,6-9H2,1H3/t11-,13-/m0/s1
Solution Type Neat
SMILES CN1CC[C@H]([C@H](CO)C1)c2ccc(F)cc2
IUPAC Name [(3S,4R)-4-(4-fluorophenyl)-1-methylpiperidin-3-yl]methanol
Molecular Weight (g/mol) 223.29
Quantity 4 x 25 mg
Formula Weight 223.1372 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.