Learn More
(3S,4R)-4-(4-Fluorophenyl)-1-methyl-3-piperidinemethanol, 98%, Thermo Scientific™
Product Code. 11317609
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11317609 25 g 25g
11307609 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11317609 Supplier Thermo Scientific Alfa Aesar Supplier No. H28137.14
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 105812-81-5
Molecular Formula C13H19FNO
Molecular Weight (g/mol) 224.30
MDL Number MFCD06658161
InChI Key CXRHUYYZISIIMT-AAEUAGOBSA-O
Synonym 3s,4r-4-4-fluorophenyl-3-hydroxymethyl-1-methylpiperidine, 3s,4r-4-4-fluorophenyl-1-methyl-3-piperidinemethanol, paroxol, unii-idr7471br7, 3s,4r-4-4-fluorophenyl-1-methylpiperidin-3-yl methanol, 3s-4-4'-fluorophenyl-3-hydroxymethyl-1-methylpiperidine, 3s,4r-4-4-fluoro-phenyl-1-methyl-piperidin-3-yl-methanol, 3-piperidinemethanol, 4-4-fluorophenyl-1-methyl-, 3s,4r, trans-4-4-fluorophenyl-3-hydroxymethyl-1-methyl-piperidine
PubChem CID 2734218
IUPAC Name [(3S,4R)-4-(4-fluorophenyl)-1-methylpiperidin-3-yl]methanol
SMILES C[NH+]1CC[C@H]([C@H](CO)C1)C1=CC=C(F)C=C1
Specifications
Melting Point 93°C to 98°C
Quantity 25 g
UN Number UN3077
Optical Rotation −34° (c=1 in Methanol)
Formula Weight 223.29
Percent Purity 98%
Chemical Name or Material (3S,4R)-4-(4-Fluorophenyl)-1-methyl-3-piperidinemethanol