missing translation for 'onlineSavingsMsg'
Learn More

(-)-(3S)-3-Hydroxy Quinine-vinyl-d3, TRC

Product Code. 30404943
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30404943 10 mg 10mg
30406337 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30404943 Supplier Toronto Research Chemicals Supplier No. TRCH95323710MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (-)-(3S)-3-Hydroxy Quinine-d3 (vinyl-d3)
Molecular Formula C20 H21 D3 N2 O3
Quantity 10 mg
Synonym (-)-(3S)-3-Hydroxy Quinine-vinyl-d3, (3S)-3-Hydroxyquinine-d3, (3S)-Hydroxyquinine-d3, 3-Hydroxyquinine-d3
InChI Formula InChI=1S/C20H24N2O3/c1-3-20(24)12-22-9-7-13(20)10-18(22)19(23)15-6-8-21-17-5-4-14(25-2)11-16(15)17/h3-6,8,11,13,18-19,23-24H,1,7,9-10,12H2,2H3/t13-,18-,19+,20+/m0/s1/i1D2,3D
SMILES O[C@]1(/C([2H])=C([2H])/[2H])C[N@]2[C@@]([C@H](O)C3=CC=NC4=CC=C(OC)C=C43)([H])C[C@@H]1CC2
IUPAC Name (1S,3S,4S,6S)-6-[(R)-hydroxy-(6-methoxy-4-quinolyl)methyl]-3-(1,2,2-trideuteriovinyl)quinuclidin-3-ol
Molecular Weight (g/mol) 343.43465
Formula Weight 343.1975
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.