missing translation for 'onlineSavingsMsg'
Learn More

(3R,5R)-7-[2-(4-Fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl]-5-hydroxy-3-methoxyheptanoic Acid Calcium Salt (3-O-Methylatorvastatin Calcium Salt), Mikromol™

Product Code. 30051785
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30051785 25 mg 25mg
30051382 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30051785 Supplier Mikromol Supplier No. MM0400.120025
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

(3R,5R)-7-[2-(4-Fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl]-5-hydroxy-3-methoxyheptanoic Acid Calcium Salt (3-O-Methylatorvastatin Calcium Salt)
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (3R,5R)-7-[2-(4-Fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl]-5-hydroxy-3-methoxyheptanoic Acid Calcium Salt (3-O-Methylatorvastatin Calcium Salt)
CAS 887196-29-4
Analyte or Component Names (3R,5R)-7-[2-(4-Fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl]-5-hydroxy-3-methoxyheptanoic Acid Calcium Salt (3-O-Methylatorvastatin Calcium Salt)
Shipping Condition Room Temperature
Molecular Formula 2 C34 H36 F N2 O5 . Ca
Certifications/Compliance ISO 9001
Synonym Atorvastatin Calcium Trihydrate Imp. G (EP), Atorvastatin Imp. G (EP), (3R,5R)-7-[2-(4-Fluorophenyl)-5-(1-methylethyl)-3-phenyl-4-(phenylcarbamoyl)-1H-pyrrol-1-yl]-5-hydroxy-3-methoxyheptanoic Acid Calcium Salt, Atorvastatin Calcium Trihydrate Imp. G (EP) as Calcium Salt, 3-O-Methylatorvastatin Calcium Salt, Atorvastatin Calcium Trihydrate Impurity G as Calcium Salt, Atorvastatin Impurity G as Calcium Salt
InChI Formula InChI=1S/2C34H37FN2O5.Ca/c2*1-22(2)32-31(34(41)36-26-12-8-5-9-13-26)30(23-10-6-4-7-11-23)33(24-14-16-25(35)17-15-24)37(32)19-18-27(38)20-28(42-3)21-29(39)40;/h2*4-17,22,27-28,38H,18-21H2,1-3H3,(H,36,41)(H,39,40);/q;;+2/p-2/t2*27-,28-;/m11./s1
SMILES [Ca+2].CO[C@H](C[C@H](O)CCn1c(C(C)C)c(C(=O)Nc2ccccc2)c(c3ccccc3)c1c4ccc(F)cc4)CC(=O)[O-].CO[C@H](C[C@H](O)CCn5c(C(C)C)c(C(=O)Nc6ccccc6)c(c7ccccc7)c5c8ccc(F)cc8)CC(=O)[O-]
IUPAC Name calcium;(3R,5R)-7-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]-5-hydroxy-3-methoxyheptanoate
Molecular Weight (g/mol) 1183.39
Quantity 25 mg
Formula Weight 1182.4842 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.