Learn More
3M™ Versaflo™ M-400 Series PAPR Accessory, Jaw Gasket
Product Code. 15780068 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
Versaflo M-400 Series PAPR System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15780068 Versaflo M-400 Series PAPR System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15780068 Supplier 3M™ Supplier No. 7100063885
  • ONLINE EXCLUSIVE PRICE
    49.00 EUR
    33.14 EUR / Pack of 1
    Save 15.86 EUR
    32% Off
Estimated Shipment: 26-01-2027
Jaw gasket is a replacement part for Versaflo™ M-400 Series helmets.
  • 3M Catalog# M-441
TRUSTED_SUSTAINABILITY
Specifications
Color Gray
Disposable Reusable
Product Type Gasket
For Use With (Application) Cleaning, Electrical, Facility Maintenance, Painting, Sanding, Welding
For Use With (Equipment) Versaflo M-400 Series PAPR System
Model M-441