Learn More
3M™ 6000 / 7800 Series Full Facepiece Accessory, Inhalation Port Gasket
Product Code. 15195724 Shop All 3M Products
Change view
Click to view available options
For Use With (Equipment):
6000, 7800 Series Full Facepiece
Unit Size:
Pack of 20
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
15195724 6000, 7800 Series Full Facepiece Pack of 20
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15195724 Supplier 3M™ Supplier No. 7000029669
  • ONLINE EXCLUSIVE PRICE
    53.00 EUR
    29.63 EUR / Pack of 20
    Save 23.37 EUR
    44% Off
View all promotions
 Restricted Item
Estimated Shipment: 19-11-2026
Inhalation port gasket for use with full-facepiece respirators.
  • These safety products must be used in accordance with the user instructions, warnings and limitations accompanying each product.
  • 3M Catalog# 6895
TRUSTED_SUSTAINABILITY
Specifications
Color Orange
Product Type Gasket
For Use With (Application) Automotive, metal production, fabrication, pharmaceutical, transportation, military maintenance, construction, food and beverage manufacturing, grinding, sanding, welding
For Use With (Equipment) 6000, 7800 Series Full Facepiece
Model 6895