missing translation for 'onlineSavingsMsg'
Learn More

(3E,7E)-4,8,12-Trimethyl-3,7,11-tridecatrienoic Acid, TRC

Product Code. 30456252
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30456252 10 mg 10mg
30453248 50 mg 50mg
30457074 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30456252 Supplier Toronto Research Chemicals Supplier No. TRCT79815510MG
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 462-66-8
Molecular Formula C16H26O2
Molecular Weight (g/mol) 250.38
Synonym Farnesylcarboxylic Acid, Homofarnesenic Acid, Homofarnesylic Acid, (E,E)-Homofarnesic Acid, (3E,7E)-4,8,12-Trimethyl-3,7,11-tridecanetrienolic Acid
IUPAC Name (3E,7E)-4,8,12-trimethyltrideca-3,7,11-trienoic acid
SMILES CC(=CCC\C(=C\CC\C(=C\CC(=O)O)\C)\C)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C16H26O2/c1-13(2)7-5-8-14(3)9-6-10-15(4)11-12-16(17)18/h7,9,11H,5-6,8,10,12H2,1-4H3,(H,17,18)/b14-9+,15-11+
Formula Weight 250.1933
Chemical Name or Material (3E,7E)-4,8,12-Trimethyl-3,7,11-tridecatrienoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.