missing translation for 'onlineSavingsMsg'
Learn More

3Beta,5Alpha-Cholic Acid-d5, TRC

Product Code. 30349079
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349079 1 mg 1mg
30354267 2.5 mg 2.5mg
30363827 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30349079 Supplier Toronto Research Chemicals Supplier No. TRCC4326171MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3beta,5alpha-Cholic Acid-d5
Molecular Formula C24H35D5O5
Quantity 1 mg
Synonym (3β,5α,7α,12α)-3,7,12-Trihydroxycholan-24-oic Acid-d5, 3β,7α,12α-Trihydroxy-5α-cholan-24-oic Acid-d5
InChI Formula InChI=1S/C24H40O5/c1-13(4-7-21(28)29)16-5-6-17-22-18(12-20(27)24(16,17)3)23(2)9-8-15(25)10-14(23)11-19(22)26/h13-20,22,25-27H,4-12H2,1-3H3,(H,28,29)/t13-,14-,15+,16-,17+,18?,19-,20+,22?,23+,24-/m1/s1/i8D2,10D2,15D
SMILES O[C@@]1([2H])C([2H])([2H])C[C@@]2(C)[C@](C[C@@H](O)C3C2C[C@H](O)[C@@]4(C)[C@@]3([H])CC[C@@H]4[C@@H](CCC(O)=O)C)([H])C1([2H])[2H]
IUPAC Name (4R)-4-((3S,5R,7R,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl-2,2,3,4,4-d5)pentanoic acid
Molecular Weight (g/mol) 413.6
Formula Weight 413.32
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.