Learn More
(3Beta)-3-(Acetyloxy)pregna-5,14,16-trien-20-one, TRC
Specifications
Specifications
| Chemical Name or Material | (3beta)-3-(Acetyloxy)pregna-5,14,16-trien-20-one |
| CAS | 2453-93-2 |
| Molecular Formula | C23H30O3 |
| Quantity | 5 mg |
| Synonym | (3b)-3-(Acetyloxy)pregna-5,14,16-trien-20-one, 3β-hydroxypregna-5,14,16-trien-20-one |
| InChI Formula | InChI=1S/C23H30O3/c1-14(24)19-7-8-20-18-6-5-16-13-17(26-15(2)25)9-11-22(16,3)21(18)10-12-23(19,20)4/h5,7-8,17-18,21H,6,9-13H2,1-4H3/t17-,18-,21?,22-,23+/m0/s1 |
| SMILES | [H][C@@]12CC=C3C[C@@H](OC(C)=O)CC[C@]3(C)C1CC[C@@]4(C)C2=CC=C4C(C)=O |
| IUPAC Name | (3S,8R,10R,13S)-17-acetyl-10,13-dimethyl-2,3,4,7,8,9,10,11,12,13-decahydro-1H-cyclopenta[a]phenanthren-3-yl acetate |
| Molecular Weight (g/mol) | 354.48 |
| Formula Weight | 354.22 |