Learn More
Uvex™ 3B Chem Light Chemical Protection Coveralls
Product Code. 17767649 Shop All Uvex Products
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
17767649 X-Large Pack of 25
17737649 Small Pack of 25
17747649 Medium Pack of 25
17757649 Large Pack of 25
17777649 2X-Large Pack of 25
17787649 3X-Large Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 17767649 Supplier Uvex™ Supplier No. 8984312
  • ONLINE EXCLUSIVE PRICE
    281.00 EUR
    265.86 EUR / Pack of 25
    Save 15.14 EUR
    5% Off
View all promotions
Estimated Shipment: 16-11-2026
Disposable coveralls constructed of polypropylene SMS laminated with polyethylene film for maximum safety.
  • Light and highly flexible material for high wearer comfort
  • Liquid-tight
  • Low-noise material
  • Free from paint wetting impairment substances and silicone
  • Ideal for cleaning work
  • Middle finger loops for securing the sleeves
  • Optimal protection thanks to self-adhesive zipper flap
  • Anti-static properties
  • Protection against infectious diseases
  • Lined with skin-friendly non-woven material
  • Secure and comfortable closures thanks to elasticated bands on hood, arms and legs
  • Elasticated waistband for optimal fit
  • Two-way zip
TRUSTED_SUSTAINABILITY
Specifications
Product Type Chemical Protective Clothing
Size X-Large
Gender Unisex
Material Polypropylene
Color White, Yellow, Orange
Disposable Single-use
Waist Style Elastic
Ankle Style Elastic
Cuff Style Covered Elastic
Neck Style Hooded
Hood Type Standard
Closure Type Zipper with Self-adhesive Flap
Seam Type Taped
Sterility Non-sterile
Resistance Chemicals
Certifications/Compliance EN 14126, EN 14605 - type 3B, EN 14605 - type 4B, ISO 13982-1 - type 5B, EN 13034 - type 6B, EN 1149-5, EN 1073-2, ISO 27065 C3