Learn More
(3aS,4S,6R,6aS)-6-((R)-2,2-Dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-ol, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 14131-84-1 |
| Molecular Formula | C12H20O6 |
| MDL Number | MFCD00134206 |
| Quantity | 1 g |
| SMILES | O[C@@H]1[C@@]2([C@]([C@](O1)([C@]3(COC(C)(C)O3)[H])[H])(OC(C)(C)O2)[H])[H] |
| Molecular Weight (g/mol) | 260.28 |
| Percent Purity | 98% |
| Chemical Name or Material | (3aS,4S,6R,6aS)-6-((R)-2,2-Dimethyl-1,3-dioxolan-4-yl)-2,2-dimethyltetrahydrofuro[3,4-d][1,3]dioxol-4-ol |