Learn More
3Alpha,17,20Beta-Trihydroxy-5Beta-pregnan-11-one, TRC
Product Code. 30455318
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455318 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30455318 Supplier Toronto Research Chemicals Supplier No. TRCT7952705MG
 Restricted Item
Estimated Shipment: 10-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3Alpha,17,20Beta-Trihydroxy-5Beta-pregnan-11-one
CAS 13933-74-9
Molecular Formula C21H34O4
Quantity 5 mg
Synonym (3α,5β,20R)-3,17,20-Trihydroxyprgnan-11-one
InChI Formula InChI=1S/C21H34O4/c1-12(22)21(25)9-7-16-15-5-4-13-10-14(23)6-8-19(13,2)18(15)17(24)11-20(16,21)3/h12-16,18,22-23,25H,4-11H2,1-3H3/t12-,13-,14+,15+,16-,18-,19+,20+,21+/m1/s1
SMILES C[C@@H](O)[C@@]1(O)CC[C@@H]2[C@@H]3CC[C@@H]4C[C@@H](O)CC[C@]4(C)[C@H]3C(=O)C[C@]12C
IUPAC Name (3S,5R,8S,9S,10S,13S,14R,17R)-3,17-dihydroxy-17-[(1R)-1-hydroxyethyl]-10,13-dimethyl-2,3,4,5,6,7,8,9,12,14,15,16-dodecahydro-1H-cyclopenta[a]phenanthren-11-one
Molecular Weight (g/mol) 350.49
Formula Weight 350.2457