Learn More
Applied Biosystems™ Sequencing Standard, BigDye™ Terminator v3.1 Dx
Product Code. 15918856 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
4 Tubes
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15918856 4 Tubes 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15918856 Supplier Applied Biosystems™ Supplier No. 4404310
 Restricted Item
In Stock
Sequencing Standard, BigDye Terminator v3.1 Dx, contains four tubes, each of which contains enough DNA of a known 1200 base pair sequence prepared with BigDye™ Terminator v3.1.
Sequencing Standard, BigDye™ Terminator v3.1 Dx, contains four tubes, each of which contains enough DNA of a known 1200 base pair sequence prepared with BigDye™ Terminator v3.1 to perform one spectral calibration/performance check and/or one control sequencing run on Applied Biosystems Dx genetic analyzers.
The DNA contained in the preparation has been lyophilized to maximize stability. Note: Applied Biosystems Dx genetic analyzers and consumables are for distribution and use in specific European countries only. Not for use in the USA.
  • The BigDye™ Terminator sequencing standard kit contains four tubes, each of which contains enough DNA of a known 1200 base pair sequence prepared with BigDye™ Terminator v3.1 to perform one spectral calibration/performance check and/or one control sequencing run on the 3500 Dx Series Genetic Analyzers.
  • The DNA contained in the preparation has been lyophilized to maximize stability.
  • Note: The 3500 Dx Series Systems and consumables are for distribution and use in specific European countries only. Not for use in the USA
TRUSTED_SUSTAINABILITY
Specifications
Quantity 4 Tubes
Product Line BigDye Terminator
For Use With (Equipment) 3500xL Dx Genetic Analyzer, 3500 Dx Genetic Analyzer
Product Type Sequencing Standard
Shipping Condition Dry Ice
Content And Storage Store in freezer (-5°C to -30°C).
For In Vitro Diagnostic Use Only