Learn More
5-Bromo-4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile, 97%, BLD Pharm™
Specifications
Specifications
| CAS | 23819-87-6 |
| Molecular Formula | C8H7BrN2O |
| MDL Number | MFCD01117224 |
| Quantity | 100 g |
| SMILES | N#CC1=C(C)C(Br)=C(C)NC1=O |
| Molecular Weight (g/mol) | 227.06 |
| Percent Purity | 97% |
| Chemical Name or Material | 5-Bromo-4,6-dimethyl-2-oxo-1,2-dihydropyridine-3-carbonitrile |