Learn More
1-(4-(2-Oxo-2-(pyrrolidin-1-yl)ethyl)piperazin-1-yl)-3-(3,4,5-trimethoxyphenyl)prop-2-en-1-one maleate, 99%, BLD Pharm™
Specifications
Specifications
| CAS | 26328-04-1 |
| Molecular Formula | C26H35N3O9 |
| MDL Number | MFCD01320526 |
| Quantity | 250 mg |
| SMILES | O=C(N1CCN(CC(N2CCCC2)=O)CC1)C=CC3=CC(OC)=C(OC)C(OC)=C3.O=C(O)/C=C\C(O)=O |
| Molecular Weight (g/mol) | 533.57 |
| Percent Purity | 99% |
| Chemical Name or Material | 1-(4-(2-Oxo-2-(pyrrolidin-1-yl)ethyl)piperazin-1-yl)-3-(3,4,5-trimethoxyphenyl)prop-2-en-1-one maleate |