missing translation for 'onlineSavingsMsg'
Learn More

n-Nonanoic-9,9,9-d3 acid, TRC

Product Code. 30468004
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30468004 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30468004 Supplier Toronto Research Chemicals Supplier No. TRCN64938710MG
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 134646-27-8
Molecular Formula C9 D3 H15 O2
Molecular Weight (g/mol) 161.256
Synonym Nonanoic-9,9,9-d3 acid (9CI), Nonanoic-9,9,9-D3 acid, 1-Octanecarboxylic-8,8,8-d3 acid, n-Nonanoic acid-d3, Pelargonic acid-ω-d3, Nonanoic acid-d3
IUPAC Name 9,9,9-trideuteriononanoic acid
SMILES [2H]C([2H])([2H])CCCCCCCC(=O)O

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C9H18O2/c1-2-3-4-5-6-7-8-9(10)11/h2-8H2,1H3,(H,10,11)/i1D3
Formula Weight 161.15
Chemical Name or Material n-Nonanoic-9,9,9-d3 acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.