missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Norethisterone Enanthate, TRC
Specifications
Specifications
| Chemical Name or Material | Norethisterone Enantate (Norethindrone Enanthate) |
| CAS | 3836-23-5 |
| Molecular Formula | C27 H38 O3 |
| Quantity | 250 mg |
| Synonym | (17α)-17-[(1-oxoheptyl)oxy]-19-Norpregn-4-en-20-yn-3-one, 19-Nor-17α-pregn-4-en-20-yn-3-one','17-hydroxy-','heptanoate (6CI,7CI,8CI), Heptanoic acid','ester with 17-hydroxy-19-nor-17α-pregn-4-en-20-yn-3-one (8CI), 17α-Ethynyl-19-nortestosterone 17-heptanoate, 17α-Ethynyl-19-nortestosterone enanthate, NSC 22846, Norethindrone enanthate, Norethisterone enanthate, Norethisterone heptanoate, Norigest, Noristerat, SH 8.0393, ZK 5410 |
| InChI Formula | InChI=1S/C27H38O3/c1-4-6-7-8-9-25(29)30-27(5-2)17-15-24-23-12-10-19-18-20(28)11-13-21(19)22(23)14-16-26(24,27)3/h2,18,21-24H,4,6-17H2,1,3H3/t21-,22+,23+,24-,26-,27-/m0/s1 |
| SMILES | CCCCCCC(=O)O[C@]1(CC[C@H]2[C@@H]3CCC4=CC(=O)CC[C@@H]4[C@H]3CC[C@]12C)C#C |
| IUPAC Name | [(8R,9S,10R,13S,14S,17R)-17-ethynyl-13-methyl-3-oxo-1,2,6,7,8,9,10,11,12,14,15,16-dodecahydrocyclopenta[a]phenanthren-17-yl] heptanoate |
| Molecular Weight (g/mol) | 410.59 |
| Formula Weight | 410.2821 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?