missing translation for 'onlineSavingsMsg'
Learn More

Tegoprazan, TRC

Product Code. 30411629
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30411629 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30411629 Supplier Toronto Research Chemicals Supplier No. TRCT14503050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Tegoprazan
CAS 942195-55-3
Purity Grade Notes HPLC
Molecular Formula C20 H19 F2 N3 O3
Quantity 50 mg
Synonym Tegoprazan, 1H-Benzimidazole-5-carboxamide','7-[[(4S)-5,7-difluoro-3,4-dihydro-2H-1-benzopyran-4-yl]oxy]-N,N,2-trimethyl-, 7-[[(4S)-5,7-Difluoro-3,4-dihydro-2H-1-benzopyran-4-yl]oxy]-N,N,2-trimethyl-1H-benzimidazole-5-carboxamide, Tegoprazan
InChI Formula InChI=1S/C20H19F2N3O3/c1-10-23-14-6-11(20(26)25(2)3)7-17(19(14)24-10)28-15-4-5-27-16-9-12(21)8-13(22)18(15)16/h6-9,15H,4-5H2,1-3H3,(H,23,24)/t15-/m0/s1
SMILES CN(C)C(=O)c1cc(O[C@H]2CCOc3cc(F)cc(F)c23)c4[nH]c(C)nc4c1
IUPAC Name 7-[[(4S)-5,7-difluoro-3,4-dihydro-2H-chromen-4-yl]oxy]-N,N,2-trimethyl-1H-benzimidazole-5-carboxamide
Molecular Weight (g/mol) 387.38
Formula Weight 387.139
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.