missing translation for 'onlineSavingsMsg'
Learn More

Amplicaine-d5, TRC

Product Code. 30331155
Change view
Click to view available options
Quantity:
5 mg
50 mg
Unit Size:
50mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30331155 5 mg 5mg
30331154 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30331155 Supplier Toronto Research Chemicals Supplier No. TRCA6342625MG
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Amplicaine-d5
Molecular Formula C14 D5 H17 N2 O
Quantity 5 mg
Synonym Octacaine-d5, 3-(Diethylamino)butyranilide-d5, 3-(Diethylamino)-N-phenyl-butanamide-d5
InChI Formula InChI=1S/C14H22N2O/c1-4-16(5-2)12(3)11-14(17)15-13-9-7-6-8-10-13/h6-10,12H,4-5,11H2,1-3H3,(H,15,17)/i6D,7D,8D,9D,10D
SMILES [2H]c1c([2H])c([2H])c(NC(=O)CC(C)N(CC)CC)c([2H])c1[2H]
IUPAC Name 3-(diethylamino)-N-(2,3,4,5,6-pentadeuteriophenyl)butanamide
Molecular Weight (g/mol) 239.368
Formula Weight 239.205
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.