missing translation for 'onlineSavingsMsg'
Learn More

Atorvastatin Allyl Ester, TRC

Product Code. 30329435
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30329435 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329435 Supplier Toronto Research Chemicals Supplier No. TRCA79171025MG
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Atorvastatin Allyl Ester
CAS 915092-85-2
Molecular Formula C36H39FN2O5
Quantity 25 mg
Synonym 2-(4-Fluorophenyl)-beta,delta-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrole-1-heptanoic Acid 2-Propenyl Ester, (betaR,deltaR)-2-(4-Fluorophenyl)-beta,delta-dihydroxy-5-(1-methylethyl)-3-phenyl-4-[(phenylamino)carbonyl]-1H-pyrrole-1-heptanoic Acid 2-Propen-1-yl Ester
InChI Formula InChI=1S/C36H39FN2O5/c1-4-21-44-31(42)23-30(41)22-29(40)19-20-39-34(24(2)3)33(36(43)38-28-13-9-6-10-14-28)32(25-11-7-5-8-12-25)35(39)26-15-17-27(37)18-16-26/h4-18,24,29-30,40-41H,1,19-23H2,2-3H3,(H,38,43)/t29-,30-/m1/s1
SMILES CC(C)c1c(C(=O)Nc2ccccc2)c(c3ccccc3)c(c4ccc(F)cc4)n1CC[C@@H](O)C[C@@H](O)CC(=O)OCC=C
IUPAC Name prop-2-enyl (3R,5R)-7-[2-(4-fluorophenyl)-3-phenyl-4-(phenylcarbamoyl)-5-propan-2-ylpyrrol-1-yl]-3,5-dihydroxyheptanoate
Molecular Weight (g/mol) 598.7
Formula Weight 598.2843
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.