missing translation for 'onlineSavingsMsg'
Learn More

Afatinib Impurity AFT-8, TRC

Product Code. 30327028
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30327028 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30327028 Supplier Toronto Research Chemicals Supplier No. TRCA35533510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Afatinib Impurity AFT-8
CAS 2323570-72-3
Molecular Formula C24 H27 Cl F N5 O4
Quantity 10 mg
Synonym N-[4-((3-Chloro-4-fluorophenyl)amino)-7-(((S)-tetrahydrofuran-3-yl)oxy)quinazolin-6-yl)-4-(dimethylamino)-3-hydroxybutenamide
InChI Formula InChI=1S/C24H27ClFN5O4/c1-31(2)11-15(32)8-23(33)30-21-9-17-20(10-22(21)35-16-5-6-34-12-16)27-13-28-24(17)29-14-3-4-19(26)18(25)7-14/h3-4,7,9-10,13,15-16,32H,5-6,8,11-12H2,1-2H3,(H,30,33)(H,27,28,29)/t15?,16-/m0/s1
SMILES CN(C)CC(O)CC(=O)Nc1cc2c(Nc3ccc(F)c(Cl)c3)ncnc2cc1O[C@H]4CCOC4
IUPAC Name N-[4-(3-chloro-4-fluoroanilino)-7-[(3S)-oxolan-3-yl]oxyquinazolin-6-yl]-4-(dimethylamino)-3-hydroxybutanamide
Molecular Weight (g/mol) 503.954
Formula Weight 503.174
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.