missing translation for 'onlineSavingsMsg'
Learn More

(2R)-3-Amino-2-fluoropropanoic Acid-13C3 >90% (up to 10% inorganics), TRC

Product Code. 30324974
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30324974 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30324974 Supplier Toronto Research Chemicals Supplier No. TRCA6096325MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (2R)-3-Amino-2-fluoropropanoic Acid-13C3
CAS 1217608-72-4
Molecular Formula 13C3H6FNO2
Quantity 5 mg
Synonym (R)-α-Fluoro-β-alanine-13C3, (R)-3-Amino-2-fluoropropanoic Acid-13C3
InChI Formula InChI=1S/C3H6FNO2/c4-2(1-5)3(6)7/h2H,1,5H2,(H,6,7)/t2-/m1/s1/i1+1,2+1,3+1
SMILES N[13CH2][13C@](F)([H])[13C](O)=O
IUPAC Name (2R)-3-amino-2-fluoro(1,2,3-13C3)propanoic acid
Molecular Weight (g/mol) 110.06
Formula Weight 110.0483
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.