missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Trifluoromethyl)-1H-pyrazole-5-carboxylic Acid Potassium, TRC
Specifications
Specifications
| Chemical Name or Material | 3-(Trifluoromethyl)-1H-pyrazole-5-carboxylic Acid Potassium |
| Analyte or Component Names | 3-(Trifluoromethyl)-1H-pyrazole-5-carboxylic Acid Potassium |
| Shipping Condition | Room Temperature |
| Molecular Formula | C5 H3 N2 O2 F3 .xK |
| InChI Formula | InChI=1S/C5H3F3N2O2.K/c6-5(7,8)3-1-2(4(11)12)9-10-3;/h1H,(H,9,10)(H,11,12); |
| Solution Type | Neat |
| SMILES | O=C(O)C1=CC(C(F)(F)F)=NN1.[xK] |
| IUPAC Name | 3-(trifluoromethyl)-1H-pyrazole-5-carboxylic acid potassium |
| Industry Type | Pharmaceutical |
| Molecular Weight (g/mol) | 219.18 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?