missing translation for 'onlineSavingsMsg'
Learn More

3-(trifluoromethoxy)benzylamine, 98%, Thermo Scientific™

Product Code. p-3211765 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10708113 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10708113 Supplier Thermo Scientific Acros Supplier No. 375840010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93071-75-1
MDL Number MFCD00061267
InChI Key TUPUHSXMDIWJQT-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy benzylamine, 3-trifluoromethoxy phenyl methanamine, 3-trifluoromethoxy benzyl amine, 3-trifluormethoxybenzylamine, m-trifluoromethoxybenzylamine, 3-trifluoromethoxybenzylamine, benzenemethanamine, 3-trifluoromethoxy, 1-3-trifluoromethoxy phenyl methanamine, 3-trifluoromethoxy phenyl methylamine
PubChem CID 145264
IUPAC Name [3-(trifluoromethoxy)phenyl]methanamine
SMILES C1=CC(=CC(=C1)OC(F)(F)F)CN

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H8F3NO
Linear Formula CF3OC6H4CH2NH2
Refractive Index 1.4510 to 1.4540
Quantity 1 g
Molecular Weight (g/mol) 191.15
Formula Weight 191.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 3-(Trifluoromethoxy)benzylamine
Show More Show Less
compliance-icons
Product Identifier
  • 3-(Trifluoromethoxy)benzylamine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.