missing translation for 'onlineSavingsMsg'
Learn More

3-(Trifluoromethoxy)benzylamine, 96%, Thermo Scientific™

Product Code. 11494287
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11494287 1 g 1g
11404297 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11494287 Supplier Thermo Scientific Alfa Aesar Supplier No. B20380.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93071-75-1
Molecular Formula C8H8F3NO
Molecular Weight (g/mol) 191.153
MDL Number MFCD00061267
InChI Key TUPUHSXMDIWJQT-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy benzylamine, 3-trifluoromethoxy phenyl methanamine, 3-trifluoromethoxy benzyl amine, 3-trifluormethoxybenzylamine, m-trifluoromethoxybenzylamine, 3-trifluoromethoxybenzylamine, benzenemethanamine, 3-trifluoromethoxy, 1-3-trifluoromethoxy phenyl methanamine, 3-trifluoromethoxy phenyl methylamine
PubChem CID 145264
IUPAC Name [3-(trifluoromethoxy)phenyl]methanamine
SMILES C1=CC(=CC(=C1)OC(F)(F)F)CN

Specifications

Refractive Index 1.4525
Quantity 1 g
UN Number UN2735
Beilstein 8763246
Sensitivity Air Sensitive
Formula Weight 191.15
Percent Purity 96%
Chemical Name or Material 3-(Trifluoromethoxy)benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.