Learn More
3-Thiophenemethanol, 97%
Product Code. 11311447
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11311447 5 g 5g
11321447 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11311447 Supplier Thermo Scientific Alfa Aesar Supplier No. L01935.06
Request Bulk or Custom Format
CAS: 71637-34-8 | C5H6OS | 114.16 g/mol
3-Thiophenemethanol was used in the preparation of 3-substituted thiophene conducting copolymers which has potential applications in electrochromic displays.1 It was also used in the synthesis of 4-(thiophene-3-ylmethoxy)phthalonitrile
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Thiophenemethanol was used in the preparation of 3-substituted thiophene conducting copolymers which has potential applications in electrochromic displays.1 It was also used in the synthesis of 4-(thiophene-3-ylmethoxy)phthalonitrile
Solubility
Slightly soluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 71637-34-8
Molecular Formula C5H6OS
Molecular Weight (g/mol) 114.16
MDL Number MFCD00014534
InChI Key BOWIFWCBNWWZOG-UHFFFAOYSA-N
Synonym 3-thiophenemethanol, 3-thienylmethanol, thiophene-3-methanol, 3-thiophene methanol, 3-hydroxymethylthiophene, thien-3-ylmethanol, thiophen-3-yl methanol, 3-thienylmethan-1-ol, 3-hydroxymethyl thiophene, 3-thiophenylmethanol
PubChem CID 123570
SMILES OCC1=CSC=C1
Specifications
Density 1.21
Boiling Point 86°C to 88°C (10 mmHg)
Flash Point 110°C (230°F)
Refractive Index 1.565
Quantity 5 g
Beilstein 106460
Solubility Information Slightly soluble in water.
IUPAC Name thiophen-3-ylmethanol
Formula Weight 114.17
Percent Purity 98%
Chemical Name or Material 3-Thiophenemethanol