Learn More
3-tert-Butyl-2-hydroxybenzaldehyde, TRC
Product Code. 30410225
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30410225 5 g
30418259 1 g
30415740 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30410225 Supplier Toronto Research Chemicals Supplier No. TRCT1357155G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 24623-65-2
Molecular Formula C11H14O2
Molecular Weight (g/mol) 178.22
Synonym 3-(1,1-Dimethylethyl)-2-hydroxybenzaldehyde, 2-Hydroxy-3-tert-butylbenzaldehyde, 3-(tert-Butyl)-2-hydroxybenzaldehyde, 3-t-Butyl-2-hydroxybenzaldehyde, 3-tert-Butylsalicylaldehyde
IUPAC Name 3-tert-butyl-2-hydroxybenzaldehyde
SMILES CC(C)(C)c1cccc(C=O)c1O
Specifications
Quantity 5 g
InChI Formula InChI=1S/C11H14O2/c1-11(2,3)9-6-4-5-8(7-12)10(9)13/h4-7,13H,1-3H3
Formula Weight 178.0994
Chemical Name or Material 3-tert-Butyl-2-hydroxybenzaldehyde