missing translation for 'onlineSavingsMsg'
Learn More

3-Pyrimidin-2-ylbenzaldehyde, 97%, Thermo Scientific™

Product Code. 10643065
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10643065 1 g 1g
10442104 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10643065 Supplier Thermo Scientific Maybridge Supplier No. CC56304DA
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 263349-22-0
Molecular Formula C11H8N2O
Molecular Weight (g/mol) 184.20
MDL Number MFCD08056283
InChI Key ZBAZYPXXIVHUFO-UHFFFAOYSA-N
Synonym 3-pyrimidin-2-yl benzaldehyde, 2-3-formylphenyl pyrimidine, 3-2-pyrimidinyl benzaldehyde, benzaldehyde,3-2-pyrimidinyl, benzaldehyde, 3-2-pyrimidinyl, benzaldehyde, 3-2-pyrimidinyl-9ci
PubChem CID 22630818
SMILES O=CC1=CC=CC(=C1)C1=NC=CC=N1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Quantity 1 g
IUPAC Name 3-(pyrimidin-2-yl)benzaldehyde
Formula Weight 184.2
Percent Purity 97%
Chemical Name or Material 3-Pyrimidin-2-ylbenzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.