Learn More
3-Phenylacrylonitrile, TRC
Product Code. 30008706
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30008706 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30008706 Supplier Toronto Research Chemicals Supplier No. TRCP3340005G
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
CAS 4360-47-8
Molecular Formula C9 H7 N
Quantity 5 g
Synonym Cinnamonitrile, 1-Cyano-2-phenylethene, 1-Cyano-2-phenylethylene, 3-Phenyl-2-propenenitrile, 3-Phenylpropenenitrile, NSC 42118, NSC 49137, Styryl cyanide, β-Cyanostyrene, β-Phenylacrylonitrile
InChI Formula InChI=1S/C9H7N/c10-8-4-7-9-5-2-1-3-6-9/h1-7H
SMILES N#CC=Cc1ccccc1
IUPAC Name 3-phenylprop-2-enenitrile
Molecular Weight (g/mol) 129.158
Formula Weight 129.0578
Chemical Name or Material 3-Phenylacrylonitrile