missing translation for 'onlineSavingsMsg'
Learn More

3-Phenanthrol, TRC

Product Code. 30012729
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012729 5 mg 5mg
30012236 2.5 mg 2.5mg
30009723 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012729 Supplier Toronto Research Chemicals Supplier No. TRCP2950105MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 605-87-8
Molecular Formula C14 H10 O
Molecular Weight (g/mol) 194.23
Synonym 3-Phenanthrenol, 3-Hydroxyphenanthrene, NSC 30984;
IUPAC Name phenanthren-3-ol
SMILES Oc1ccc2ccc3ccccc3c2c1

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C14H10O/c15-12-8-7-11-6-5-10-3-1-2-4-13(10)14(11)9-12/h1-9,15H
Formula Weight 194.0732
Percent Purity >95%
Chemical Name or Material 3-Hydroxyphenanthrene
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.