Learn More
3-Phenanthrol, TRC
Product Code. 30012236
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
2.5mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012236 2.5 mg 2.5mg
30012729 5 mg 5mg
30009723 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30012236 Supplier Toronto Research Chemicals Supplier No. TRCP2950102.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 605-87-8
Molecular Formula C14 H10 O
Molecular Weight (g/mol) 194.23
Synonym 3-Phenanthrenol, 3-Hydroxyphenanthrene, NSC 30984;
IUPAC Name phenanthren-3-ol
SMILES Oc1ccc2ccc3ccccc3c2c1
Specifications
Quantity 2.5 mg
InChI Formula InChI=1S/C14H10O/c15-12-8-7-11-6-5-10-3-1-2-4-13(10)14(11)9-12/h1-9,15H
Formula Weight 194.0732
Percent Purity >95%
Chemical Name or Material 3-Hydroxyphenanthrene
Grade HPLC